2-Thiophenboronsäure

Lieferant: Molekula

Synonyme: 2-Thienylboronsäure, Thiophen-2-boronsäure

M36681289
MOLEM36681289EA 0 EUR
MOLEM36681289
2-Thiophenboronsäure
2-Thiopheneboronic acid
Formel: C₄H₅BO₂S
Molecular Weight: 127,96 g/mol
Schmelzpunkt: 146…147 °C
MDL: MFCD00151850
CAS-Nummer: 6165-68-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR