N-Fmoc-L-valin

Lieferant: Molekula

M36953221
MOLEM36953221EA 0 EUR
MOLEM36953221
N-Fmoc-L-valin
N-Fmoc-L-valin
Formel: C₂₀H₂₁NO₄
Molecular Weight: 339.4 g/mol
MDL: MFCD00037124
CAS-Nummer: 68858-20-8
EINECS: 272-515-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR