4-(Methylthio)phenylboronsäure

Lieferant: Molekula

Synonyme: 4-(Methylthio)benzeneboronic acid, 4-Thioanisoleboronic acid

M37001741
MOLEM37001741EA 0 EUR
MOLEM37001741
4-(Methylthio)phenylboronsäure
4-(Methylthio)phenylboronsäure
Formel: C₇H₉BO₂S
Molecular Weight: 168,02 g/mol
MDL: MFCD00093410
CAS-Nummer: 98546-51-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR