5-Chlor-2-methylbenzoxazol
Lieferant: Molekula
Synonyme:
5-Chloro-2-methylbenzo[d]oxazole
M37075484
MOLEM37075484EA
0
EUR
MOLEM37075484
5-Chlor-2-methylbenzoxazol
5-Chloro-2-methylbenzoxazole
Formel:
C₈H₆ClNO Molecular Weight: 167,5900 g/mol |
MDL:
MFCD00022850 CAS-Nummer: 19219-99-9 EINECS: 242-888-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...