Indol-3-carbonitril

Lieferant: Molekula

M37090532
MOLEM37090532EA 0 EUR
MOLEM37090532
Indol-3-carbonitril
Indol-3-carbonitril
Formel: C₉H₆N₂
Siedepunkt: 350 °C (1013 hPa)
MDL: MFCD00022717
CAS-Nummer: 5457-28-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR