(±)-Cbz-α-phosphonoglycintrimethylester
Lieferant: Molekula
Synonyme:
N-Benzyloxycarbonyl-2-phosphonoglycine trimethyl ester, (±)-Benzyloxycarbonyl-α-phosphonoglycine trimethyl ester, (±)-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl)acetate, Methyl 2-{[(benzyloxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetate, Methyl 2-(((benzyloxy)carbonyl)amino)-2-(dimethoxyphosphoryl)acetate
M37339344
MOLEM37339344EA
0
EUR
MOLEM37339344
(±)-Cbz-α-phosphonoglycintrimethylester
(±)-Cbz-α-phosphonoglycintrimethylester
Formel:
C₁₃H₁₈NO₇P Molecular Weight: 331,27 g/mol |
MDL:
MFCD00043304 CAS-Nummer: 88568-95-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...