α-Chlor-3-nitrotoluol

Lieferant: Molekula

Synonyme: 3-Nitrobenzylchlorid, m-Nitrobenzylchlorid, 1-(Chlormethyl)-3-nitrobenzol, α-Chlor-m-nitrotoluol

M37384152
MOLEM37384152EA 0 EUR
MOLEM37384152
α-Chlor-3-nitrotoluol
α-Chlor-3-nitrotoluol
Formel: C₇H₆ClNO₂
MDL: MFCD00007272
CAS-Nummer: 619-23-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR