Diethyl-3-oxoglutarat
Lieferant: Molekula
Synonyme:
Diethyl 1,3-acetonedicarboxylate, Diethyl acetone-1,3-dicarboxylate
M37576934
MOLEM37576934EA
0
EUR
MOLEM37576934
Diethyl-3-oxoglutarat
Diethyl-3-oxoglutarat
Formel:
C₉H₁₄O₅ Molecular Weight: 202,21 g/mol |
MDL:
MFCD00009200 CAS-Nummer: 105-50-0 EINECS: 203-302-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...