6-Hydroxyindol

Lieferant: Molekula
Gefahr

Synonyme: IMEXINE OBA, Indol-6-ol, 6-Hydroxy-1H-indol, 6-Indolol, 1H-Indol-6-ol

M37611894
MOLEM37611894EA 0 EUR
MOLEM37611894
6-Hydroxyindol
6-Hydroxyindol
Formel: C₈H₇NO
Molecular Weight: 133,15 g/mol
MDL: MFCD00152101
CAS-Nummer: 2380-86-1
EINECS: 417-020-4
UN: 3077
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR