Thiazolidin-2-carbonsäure

Lieferant: Molekula

Synonyme: 1,3-Thiazolidine-2-carboxylic acid, 2-Carboxy-1,3-thiazolidine, 1,3-Thiazolane-2-carboxylic acid

M37939444
MOLEM37939444EA 0 EUR
MOLEM37939444
Thiazolidin-2-carbonsäure
Thiazolidin-2-carbonsäure
Formel: C₄H₇NO₂S
Molecular Weight: 133,169 g/mol
MDL: MFCD00068005
CAS-Nummer: 65126-70-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR