ABTS (Diammonium-2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazol-6-sulfonat])

Lieferant: Molekula
Achtung

Synonyme: ABTS, Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulphonate), 2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulphonic acid) diammonium salt

M38060073
MOLEM38060073EA 0 EUR
MOLEM38060073
ABTS (Diammonium-2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazol-6-sulfonat])
ABTS (Diammonium-2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazol-6-sulfonat])
Formel: C₁₈H₂₄N₆O₆S₄
Molecular Weight: 548,68 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00010404
CAS-Nummer: 30931-67-0
EINECS: 250-396-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR