2,4-Thiazolidindion

Lieferant: Molekula

Synonyme: 1,3-Thiazolidine-2,4-dione

M38135702
MOLEM38135702EA 0 EUR
MOLEM38135702
2,4-Thiazolidindion
2,4-Thiazolidindion
Formel: C₃H₃NO₂S
Molecular Weight: 117,13 g/mol
MDL: MFCD00005478
CAS-Nummer: 2295-31-0
EINECS: 218-941-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR