2'-Desoxyguanosin-5'-triphosphat Trinatriumsalz (dGTP-Na3)

Lieferant: Molekula

Synonyme: Deoxyguanosine triphosphate trisodium salt, 2'-Deoxyguanosine-5'-triphosphate trisodium salt

M38338086
MOLEM38338086EA 0 EUR
MOLEM38338086
2'-Desoxyguanosin-5'-triphosphat Trinatriumsalz (dGTP-Na3)
2'-Desoxyguanosin-5'-triphosphat Trinatriumsalz (dGTP-Na3)
Formel: C₁₀H₁₃N₅Na₃O₁₃P₃
Molecular Weight: 573,12 g/mol
MDL: MFCD00080337
CAS-Nummer: 93919-41-6
EINECS: 300-026-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR