Dikaliumbenzol-1,3-disulfonat

Lieferant: Molekula

M38413195
MOLEM38413195EA 0 EUR
MOLEM38413195
Dikaliumbenzol-1,3-disulfonat
Dikaliumbenzol-1,3-disulfonat
Formel: C₆H₄K₂O₆S₂
Molecular Weight: 314,42 g/mol
CAS-Nummer: 5710-54-3
EINECS: 227-199-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR