2,4-Bis(trifluormethyl)benzoesäure
Lieferant: Molekula
M38500957
MOLEM38500957EA
0
EUR
MOLEM38500957
2,4-Bis(trifluormethyl)benzoesäure
2,4-Bis(trifluormethyl)benzoesäure
Formel:
C₉H₄F₆O₂ Molecular Weight: 258,12 g/mol Siedepunkt: 260 °C (1013 hPa) Schmelzpunkt: 111…112 °C |
MDL:
MFCD00042496 CAS-Nummer: 32890-87-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...