3-Thenoesäure
Lieferant: Molekula
Synonyme:
Thiophen-3-carbonsäure, 3-Thiophencarbonsäure
M38730219
MOLEM38730219EA
0
EUR
MOLEM38730219
3-Thenoesäure
3-Thenoesäure
Formel:
C₅H₄O₂S Molecular Weight: 128,15 g/mol Schmelzpunkt: 137 °C |
MDL:
MFCD00005467 CAS-Nummer: 88-13-1 EINECS: 201-802-5 Merck Index: 12,09415 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...