Bismarckbraun R

Lieferant: Molekula

M38973565
MOLEM38973565EA 0 EUR
MOLEM38973565
Bismarckbraun R
Bismarckbraun R
Formel: C₂₁H₂₄N₈·2HCl
Lagertemperatur: Raumtemperatur
MDL: MFCD00036386
CAS-Nummer: 5421-66-9
EINECS: 213-888-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR