2,6-Dichlor-3-cyan-4-picolin
Lieferant: Molekula
Synonyme:
2,6-Dichloro-4-methylnicotinonitrile, 2,6-dichloro-4-methylpyridine-3-carbonitrile, 2,6-Dichloro-3-cyano-4-methylpyridine
M39195370
MOLEM39195370EA
0
EUR
MOLEM39195370
2,6-Dichlor-3-cyan-4-picolin
2,6-Dichlor-3-cyan-4-picolin
Formel:
C₇H₄Cl₂N₂ Molecular Weight: 187,03 g/mol |
MDL:
MFCD00052633 CAS-Nummer: 875-35-4 EINECS: 212-873-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...