3-Acetylindol
Lieferant: Molekula
Synonyme:
Indol-3-ylmethylketon, 3-Indolylmethylketon, 3-Indolyl methyl ketone, 1-(3-Indolyl)-1-ethanon
M39696970
MOLEM39696970EA
0
EUR
MOLEM39696970
3-Acetylindol
3-Acetylindol
Formel:
C₁₀H₉NO Molecular Weight: 159,19 g/mol Siedepunkt: 144…145 °C (10 mmHg) Schmelzpunkt: 186…189 °C |
MDL:
MFCD00005626 CAS-Nummer: 703-80-0 EINECS: 211-875-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...