1-Phenyl-2-tetrazolin-5-thion

Lieferant: Molekula

Synonyme: 1-Phenyl-5-mercaptotetrazol, 5-Mercapto-1-phenyl-1H-tetrazol, 1-Phenyl-1H-tetrazol-5-thiol

M39882979
MOLEM39882979EA 0 EUR
MOLEM39882979
1-Phenyl-2-tetrazolin-5-thion
1-Phenyl-2-tetrazolin-5-thion
Formel: C₇H₆N₄S
Molecular Weight: 178,22 g/mol
Schmelzpunkt: 146…150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003129
CAS-Nummer: 86-93-1
EINECS: 201-710-5
UN: 1325
ADR: 4.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR