(R)-(-)-4-Phenyl-2-oxazolidinon

Lieferant: Molekula

Synonyme: (R)-4-Phenyloxazolidin-2-one

M39921672
MOLEM39921672EA 0 EUR
MOLEM39921672
(R)-(-)-4-Phenyl-2-oxazolidinon
(R)-(-)-4-Phenyl-2-oxazolidinon
Formel: C₉H₉NO₂
Molecular Weight: 163,18 g/mol
MDL: MFCD00192393
CAS-Nummer: 90319-52-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR