2-Hydroxypyridin-N-oxid

Lieferant: Molekula

Synonyme: 2-Pyridinol-1-oxide

M39979413
MOLEM39979413EA 0 EUR
MOLEM39979413
2-Hydroxypyridin-N-oxid
2-Hydroxypyridin-N-oxid
Formel: C₅H₅NO₂
Molecular Weight: 111,10 g/mol
MDL: MFCD00006195
CAS-Nummer: 13161-30-3
EINECS: 236-100-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR