5-Aminosalicylsäure
Lieferant: Molekula
Synonyme:
Mesalamine, Mesalazine, 5-Amino-2-hydroxybenzoic acid, m-Aminosalicylic acid
M40581544
MOLEM40581544EA
0
EUR
MOLEM40581544
5-Aminosalicylsäure
5-Aminosalicylsäure
Formel:
C₇H₇NO₃ Molecular Weight: 153,14 g/mol Siedepunkt: 347 °C (1013 hPa) Schmelzpunkt: 280 °C Dichte: 1,568 g/cm³ (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00007877 CAS-Nummer: 89-57-6 EINECS: 201-919-1 Merck Index: 12,05964 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...