4-(2-Pyridylazo)resorcin

Lieferant: Molekula

Synonyme: 4-(2'-Pyridylazo)resorcinol

M40621555
MOLEM40621555EA 0 EUR
MOLEM40621555
4-(2-Pyridylazo)resorcin
4-(2-Pyridylazo)resorcin
Formel: C₁₁H₉N₃O₂
Molecular Weight: 215,21 g/mol
MDL: MFCD00044237
CAS-Nummer: 1141-59-9
EINECS: 214-528-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR