Bathophenanthrolin
Lieferant: Molekula
Synonyme:
4,7-Diphenyl-1,10-phenanthrolin
M40716309
MOLEM40716309EA
0
EUR
MOLEM40716309
Bathophenanthrolin
Bathophenanthrolin
Formel:
C₂₄H₁₆N₂ Molecular Weight: 332,4 g/mol Siedepunkt: 508 °C (1013 hPa) Schmelzpunkt: 215…220 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004976 CAS-Nummer: 1662-01-7 EINECS: 216-767-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...