Methyl-5-indolcarboxylat

Lieferant: Molekula

Synonyme: Methyl 1H-indole-5-carboxylate

M41072002
MOLEM41072002EA 0 EUR
MOLEM41072002
Methyl-5-indolcarboxylat
Methyl-5-indolcarboxylat
Formel: C₁₀H₉NO₂
Molecular Weight: 175,19 g/mol
MDL: MFCD00153023
CAS-Nummer: 1011-65-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR