3-Nitrosalicylaldehyd

Lieferant: Molekula
Achtung

Synonyme: 2-Hydroxy-3-nitrobenzaldehyde

M41263630
MOLEM41263630EA 0 EUR
MOLEM41263630
3-Nitrosalicylaldehyd
3-Nitrosalicylaldehyd
Formel: C₇H₅NO₄
Molecular Weight: 167,12 g/mol
MDL: MFCD00041874
CAS-Nummer: 5274-70-4
EINECS: 226-098-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR