α,α,α,α',α',α'-Hexafluor-2,4-xylidin
Lieferant: Molekula
Synonyme:
2,4-Bis(trifluormethyl)anilin
M41479328
MOLEM41479328EA
0
EUR
MOLEM41479328
α,α,α,α',α',α'-Hexafluor-2,4-xylidin
α,α,α,α',α',α'-Hexafluor-2,4-xylidin
Formel:
C₈H₅F₆N Molecular Weight: 229,12 g/mol Siedepunkt: 50 °C (0,4 mmHg) |
MDL:
MFCD00236209 CAS-Nummer: 367-71-5 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...