5-Indolcarbaldehyd

Lieferant: Molekula

Synonyme: Indol-5-carboxaldehyd

M41628435
MOLEM41628435EA 0 EUR
MOLEM41628435
5-Indolcarbaldehyd
5-Indolcarbaldehyd
Formel: C₉H₇NO
Molecular Weight: 145,16 g/mol
MDL: MFCD02093664
CAS-Nummer: 1196-69-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR