2-Chlorphenylboronsäure

Lieferant: Molekula
Achtung

Synonyme: 2-Chlorbenzolboronsäure, Dihydroxy(2-chlorphenyl)boran, (2-Chlorphenyl)metaborsäure

M41884294
MOLEM41884294EA 0 EUR
MOLEM41884294
2-Chlorphenylboronsäure
2-Chlorphenylboronsäure
Formel: C₆H₆BClO₂
Molecular Weight: 156,38 g/mol
MDL: MFCD00674012
CAS-Nummer: 3900-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR