D(+)-Turanose

Lieferant: Molekula

Synonyme: (3S,4R,5R)-1,4,5,6-Tetrahydroxy-3-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexan-2-one, Turanose

M41949862
MOLEM41949862EA 0 EUR
MOLEM41949862
D(+)-Turanose
D(+)-Turanose
Formel: C₁₂H₂₂O₁₁
Molecular Weight: 342.30 g/mol
MDL: MFCD00006606
CAS-Nummer: 547-25-1
EINECS: 208-918-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR