(1S)-(-)-Camphansäurechlorid

Lieferant: Molekula

Synonyme: (1S)-4,7,7-Trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carbonyl chloride, (-)-Camphanic acid chloride

M42033749
MOLEM42033749EA 0 EUR
MOLEM42033749
(1S)-(-)-Camphansäurechlorid
(1S)-(-)-Camphansäurechlorid
Formel: C₁₀H₁₃ClO₃
Molecular Weight: 216,67 g/mol
MDL: MFCD00135626
CAS-Nummer: 39637-74-6
EINECS: 254-552-4
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR