3-Chlor-o-xylol

Lieferant: Molekula

Synonyme: 3-Chlor-1,2-dimethylbenzol, 1-Chlor-2,3-dimethylbenzol

M42123136
MOLEM42123136EA 0 EUR
MOLEM42123136
3-Chlor-o-xylol
3-Chlor-o-xylol
Formel: C₈H₉Cl
Molecular Weight: 140.61 g/mol
MDL: MFCD00060689
CAS-Nummer: 608-23-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR