2-(Brommethyl)chinolin

Lieferant: Molekula

M42510135
MOLEM42510135EA 0 EUR
MOLEM42510135
2-(Brommethyl)chinolin
2-(Bromomethyl)quinoline
Formel: C₁₀H₈BrN
MDL: MFCD08062399
CAS-Nummer: 5632-15-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR