N(α)-Boc-L-histidin

Lieferant: Molekula

Synonyme: Boc-His-OH

M42799369
MOLEM42799369EA 0 EUR
MOLEM42799369
N(α)-Boc-L-histidin
N(α)-Boc-L-histidin
Formel: C₁₁H₁₇N₃O₄
Molecular Weight: 255,3 g/mol
MDL: MFCD00065576
CAS-Nummer: 17791-52-5
EINECS: 241-768-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR