(4-Nitrophenyl)-α-D-galactopyranosid

Lieferant: Molekula

Synonyme: (2R,3R,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol, 4-Nitrophenyl α-D-galactopyranoside

M42887890
MOLEM42887890EA 0 EUR
MOLEM42887890
(4-Nitrophenyl)-α-D-galactopyranosid
(4-Nitrophenyl)-α-D-galactopyranosid
Formel: C₁₂H₁₅NO₈
MDL: MFCD00065050
CAS-Nummer: 7493-95-0
EINECS: 231-343-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR