Galaktit

Lieferant: Molekula

Synonyme: Dulcitol

M42994090
MOLEM42994090EA 0 EUR
MOLEM42994090
Galaktit
Galaktit
Formel: HOH₂C(CH(OH))₄CH₂OH
Molecular Weight: 182,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00064288
CAS-Nummer: 608-66-2
EINECS: 210-165-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR