(S)-(-)-Hydroxypyrrolidin

Lieferant: Molekula

Synonyme: (S)-(-)-3-Pyrrolidinol, (S)-Pyrrolidin-3-ol, (S)-(-)-3-Hydroxypyrrolidine

M43186047
MOLEM43186047EA 0 EUR
MOLEM43186047
(S)-(-)-Hydroxypyrrolidin
(S)-(-)-Hydroxypyrrolidin
Formel: C₄H₉NO
Molecular Weight: 87,12 g/mol
MDL: MFCD00192426
CAS-Nummer: 100243-39-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR