9-Fluorenon-1-carbonsäure

Lieferant: Molekula

Synonyme: 9-Oxofluoren-1-carbonsäure

M43643129
MOLEM43643129EA 0 EUR
MOLEM43643129
9-Fluorenon-1-carbonsäure
9-Fluorenon-1-carbonsäure
Formel: C₁₄H₈O₃
Molecular Weight: 224.22 g/mol
MDL: MFCD00011537
CAS-Nummer: 1573-92-8
EINECS: 216-396-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR