N,N-Dimethylglycinethylester

Lieferant: Molekula

Synonyme: Di-Me-Gly-OEt

M43660096
MOLEM43660096EA 0 EUR
MOLEM43660096
N,N-Dimethylglycinethylester
N,N-Dimethylglycinethylester
Formel: C₆H₁₃NO₂
Molecular Weight: 131,17 g/mol
MDL: MFCD00009175
CAS-Nummer: 33229-89-9
EINECS: 251-411-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR