2-Amino-6-brompyridin

Lieferant: Molekula
Achtung

Synonyme: 6-Brompyridin-2-amin, 6-Brom-2-pyridylamin

M43883839
MOLEM43883839EA 0 EUR
MOLEM43883839
2-Amino-6-brompyridin
2-Amino-6-brompyridin
Formel: C₅H₅BrN₂
Molecular Weight: 173,01 g/mol
MDL: MFCD00137843
CAS-Nummer: 19798-81-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR