4-Chlor-3-nitrobenzolsulfonylchlorid
Lieferant: Molekula
Synonyme:
4-Chlor-3-nitrobenzolsulfonsäurechlorid, 4-Chlor-3-nitrobenzolsulfochlorid, 4-Chlor-3-nitrobenzol-1-sulfonylchlorid
M43931765
MOLEM43931765EA
0
EUR
MOLEM43931765
4-Chlor-3-nitrobenzolsulfonylchlorid
4-Chlor-3-nitrobenzolsulfonylchlorid
Formel:
C₆H₃Cl₂NO₄S Molecular Weight: 256,07 g/mol Schmelzpunkt: 61,1 °C |
MDL:
MFCD00007440 CAS-Nummer: 97-08-5 EINECS: 202-558-2 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...