3-Hydroxypyridin-N-oxid

Lieferant: Molekula

M44132528
MOLEM44132528EA 0 EUR
MOLEM44132528
3-Hydroxypyridin-N-oxid
3-Hydroxypyridin-N-oxid
Formel: C₅H₅NO₂
MDL: MFCD00006200
CAS-Nummer: 6602-28-4
EINECS: 229-545-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR