(4-Nitrophenyl)-β-D-galactopyranosid
Lieferant: Molekula
Synonyme:
pNPG, 4-Nitrophenyl β-D-Galactopyranoside
M44205949
MOLEM44205949EA
0
EUR
MOLEM44205949
(4-Nitrophenyl)-β-D-galactopyranosid
(4-Nitrophenyl)-β-D-galactopyranosid
Formel:
C₁₂H₁₅NO₈ Molecular Weight: 301,25 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00063256 CAS-Nummer: 3150-24-1 EINECS: 221-584-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...