2-Mercaptoimidazol

Lieferant: Molekula

Synonyme: Imidazol-2-thiol, 2-Mercapto-1H-imidazol

M44841057
MOLEM44841057EA 0 EUR
MOLEM44841057
2-Mercaptoimidazol
2-Mercaptoimidazole
Formel: C₃H₄N₂S
Molecular Weight: 100,14 g/mol
MDL: MFCD00005188
CAS-Nummer: 872-35-5
EINECS: 212-823-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR