4-Picolylchlorid Hydrochlorid

Lieferant: Molekula

M44972750
MOLEM44972750EA 0 EUR
MOLEM44972750
4-Picolylchlorid Hydrochlorid
4-Picolylchlorid Hydrochlorid
Formel: C₆H₇Cl₂N
MDL: MFCD00012826
CAS-Nummer: 1822-51-1
EINECS: 217-350-7
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR