2,3-Dichlor-5-(trichlormethyl)pyridin

Lieferant: Molekula

M45141459-5G
MOLEM45141459-5GEA 0 EUR
MOLEM45141459-5G
2,3-Dichlor-5-(trichlormethyl)pyridin
2,3-Dichloro-5-(trichloromethyl)pyridine
Formel: C₆H₂Cl₅N
Molecular Weight: 265.3518 g/mol
MDL: MFCD03701543
CAS-Nummer: 69045-83-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR