5-Methoxytryptamin Hydrochlorid

Lieferant: Molekula

Synonyme: 2-(5-Methoxy-1H-indol-3-yl)ethanamine, O-Methylserotonin hydrochloride

M45320712
MOLEM45320712EA 0 EUR
MOLEM45320712
5-Methoxytryptamin Hydrochlorid
5-Methoxytryptamin Hydrochlorid
Formel: C₁₁H₁₅ClN₂O
MDL: MFCD00005662
CAS-Nummer: 66-83-1
EINECS: 200-637-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR