DL-Threonin

Lieferant: Molekula

M45391267
MOLEM45391267EA 0 EUR
MOLEM45391267
DL-Threonin
DL-Threonin
Formel: C₄H₉NO₃
Molecular Weight: 119,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00063722
CAS-Nummer: 80-68-2
EINECS: 201-300-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR