1,4,8,11-Tetraazacyclotetradecan

Lieferant: Molekula

Synonyme: CYCLAM

M45676081
MOLEM45676081EA 0 EUR
MOLEM45676081
1,4,8,11-Tetraazacyclotetradecan
1,4,8,11-Tetraazacyclotetradecane
Formel: C₁₀H₂₄N₄
Molecular Weight: 200,33 g/mol
MDL: MFCD00005105
CAS-Nummer: 295-37-4
EINECS: 206-039-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR